Methyl-4-(2,3-dichloro-4-pyridinyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

046832-500MG 046832-1G
101877-182EA 921.54 USD
101877-182 101877-184
Methyl-4-(2,3-dichloro-4-pyridinyl)benzenecarboxylate ≥95%
Methyl-4-(2,3-dichloro-4-pyridinyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₉Cl₂NO₂
MW: 282.13 g/mol
Melting Pt: 171…172 °C
MDL Number: MFCD12025917
CAS Number: 1160474-76-9

Order Now

Specification Test Results

Melting point 171-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR