3-Bromo-7-chloro-1H-indazole ≥95%
Supplier: Matrix Scientific
046831-500MG
046831-1G
101877-178EA
452.31
USD
101877-178
101877-180
3-Bromo-7-chloro-1H-indazole ≥95%
3-Bromo-7-chloro-1H-indazole
Hazard: Irritant
Formula:
C₇H₄BrClN₂ Melting Pt: 155-156°C |
MDL Number:
MFCD07781601 CAS Number: 885521-96-0 |
Specification Test Results
Melting point | 155-156°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...