Ethyl-5-nitro-1H-pyrazole-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 5-nitro-1H-pyrazole-3-carboxylate
046829-500MG
046829-1G
101877-174EA
747.69
USD
101877-174
101877-176
Ethyl-5-nitro-1H-pyrazole-3-carboxylate ≥95%
Ethyl-5-nitro-1H-pyrazole-3-carboxylate
Hazard: Irritant
Formula:
C₆H₇N₃O₄ MW: 185.14 g/mol Melting Pt: 148…149 °C |
MDL Number:
MFCD04971172 CAS Number: 39846-84-9 |
Specification Test Results
Melting point | 148-149°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...