Methyl-5-chloro-6-methoxynicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-chloro-6-methoxynicotinate, Methyl 3-chloro-2-methoxypyridine-5-carboxylate

046826-500MG 046826-1G
101877-162EA 375.38 USD
101877-162 101877-164
Methyl-5-chloro-6-methoxynicotinate ≥95%
Methyl-5-chloro-6-methoxynicotinate

Hazard: Irritant

Formula: C₈H₈ClNO₃
Melting Pt: 108…110 °C
MDL Number: MFCD12025914
CAS Number: 220656-93-9

Order Now

Specification Test Results

Melting point 108-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR