(5-Nitro-1H-pyrazol-3-yl)methanol ≥95%
Supplier: Matrix Scientific
046824-500MG
046824-1G
101877-154EA
818.46
USD
101877-154
101877-156
(5-Nitro-1H-pyrazol-3-yl)methanol ≥95%
(5-Nitro-1H-pyrazol-3-yl)methanol
Hazard: Irritant
Formula:
C₄H₅N₃O₃ Melting Pt: 136…137 °C |
MDL Number:
MFCD12025912 CAS Number: 1000895-25-9 |
Specification Test Results
Melting point | 136-137°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...