2,5-Difluoro-4-nitrobenzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
2,5-Difluoro-4-nitrobenzaldehyde
046823-500MG
046823-1G
101877-150EA
921.54
USD
101877-150
101877-152
2,5-Difluoro-4-nitrobenzenecarbaldehyde ≥95%
2,5-Difluoro-4-nitrobenzenecarbaldehyde
Hazard: Irritant
Formula:
C₇H₃F₂NO₃ MW: 187.1 g/mol Melting Pt: 51…53 °C |
MDL Number:
MFCD09909605 CAS Number: 1160474-72-5 |
Specification Test Results
Melting point | 51-53°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...