Methyl-2,3-dichloroisonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,3-dichloroisonicotinate

046822-500MG 046822-1G 046822-5G
101877-144EA 118.46 USD
101877-144 101877-146 101877-148
Methyl-2,3-dichloroisonicotinate ≥95%
Methyl 2,3-dichloroisonicotinate

Hazard: Irritant

Formula: C₇H₅Cl₂NO₂
MW: 206.0300 g/mol
Melting Pt: 38-39°C
MDL Number: MFCD09951946
CAS Number: 603124-78-3

Order Now

Specification Test Results

Melting point 38-39°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR