Methyl-5-amino-1H-pyrazole-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-amino-1H-pyrazole-3-carboxylate
046815-500MG
046815-1G
101877-122EA
200
USD
101877-122
101877-124
Methyl-5-amino-1H-pyrazole-3-carboxylate ≥95%
Methyl-5-amino-1H-pyrazole-3-carboxylate
Hazard: Irritant
Formula:
C₅H₇N₃O₂ MW: 141.1300 g/mol Melting Pt: 135…136 °C |
MDL Number:
MFCD08063165 CAS Number: 632365-54-9 |
Specification Test Results
Melting point | 135-136°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...