2-[(3-Chloro-5-nitro-2-pyridinyl)amino]-1-ethanol ≥95%

Supplier: Matrix Scientific

046807-500MG 046807-1G
101877-102EA 375.38 USD
101877-102 101877-104
2-[(3-Chloro-5-nitro-2-pyridinyl)amino]-1-ethanol ≥95%
2-[(3-Chloro-5-nitro-2-pyridinyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₇H₈ClN₃O₃
Melting Pt: 139…141 °C
MDL Number: MFCD12025906
CAS Number: 1160474-59-8

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR