Ethyl-6,8-dichloro-2-(trifluoromethyl)-2H-chromene-3-carboxylate ≥95%

Supplier: Matrix Scientific

046803-500MG 046803-1G
101877-086EA 375.38 USD
101877-086 101877-088
Ethyl-6,8-dichloro-2-(trifluoromethyl)-2H-chromene-3-carboxylate ≥95%
Ethyl-6,8-dichloro-2-(trifluoromethyl)-2H-chromene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₉Cl₂F₃O₃
Melting Pt: 106…108 °C
MDL Number: MFCD12025902
CAS Number: 1160474-53-2

Order Now

Specification Test Results

Melting point 106-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR