2-(4-Oxopentyl)-1H-isoindole-1,3(2H)-dione ≥95%
Supplier: Matrix Scientific
046802-1G
046802-5G
101877-082EA
106.15
USD
101877-082
101877-084
2-(4-Oxopentyl)-1H-isoindole-1,3(2H)-dione ≥95%
2-(4-Oxopentyl)-1H-isoindole-1,3(2H)-dione
Hazard: Irritant
Formula:
C₁₃H₁₃NO₃ MW: 231.25 g/mol Melting Pt: 74…75 °C |
MDL Number:
MFCD06797717 CAS Number: 3197-25-9 |
Specification Test Results
Melting point | 74-75°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...