Ethyl-6-bromo-2-(trifluoromethyl)-2H-chromene-3-carboxylate ≥95%

Supplier: Matrix Scientific

046801-1G 046801-5G
101877-076EA 553.85 USD
101877-076 101877-078
Ethyl-6-bromo-2-(trifluoromethyl)-2H-chromene-3-carboxylate ≥95%
Ethyl-6-bromo-2-(trifluoromethyl)-2H-chromene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₀BrF₃O₃
Melting Pt: 81…83 °C
MDL Number: MFCD12025901
CAS Number: 1160474-50-9

Order Now

Specification Test Results

Melting point 81-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR