6-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

046799-500MG 046799-1G
101877-064EA 292.31 USD
101877-064 101877-066
6-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
6-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₄BrF₃N₂
Melting Pt: 106…108 °C
MDL Number: MFCD11111691
CAS Number: 150780-40-8

Order Now

Specification Test Results

Melting point 106-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR