1-[4-Nitro-2-(trifluoromethyl)phenyl]-1H-pyrazole ≥95%

Supplier: Matrix Scientific

046796-500MG 046796-1G 046796-5G
101877-048EA 181.54 USD
101877-048 101877-050 101877-052
1-[4-Nitro-2-(trifluoromethyl)phenyl]-1H-pyrazole ≥95%
1-[4-Nitro-2-(trifluoromethyl)phenyl]-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₆F₃N₃O₂
Melting Pt: 45…47 °C
MDL Number: MFCD12498690
CAS Number: 1177093-03-6

Order Now

Specification Test Results

Melting point 45-47°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR