2-[5-Methoxy-2-nitro-4-(trifluoromethyl)phenyl]acetonitrile ≥97%

Supplier: Matrix Scientific

046795-500MG 046795-1G 046795-5G
101877-042EA 315.38 USD
101877-042 101877-044 101877-046
2-[5-Methoxy-2-nitro-4-(trifluoromethyl)phenyl]acetonitrile ≥97%
2-[5-Methoxy-2-nitro-4-(trifluoromethyl)phenyl]acetonitrile

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O₃
MW: 260.17 g/mol
Melting Pt: 105…106 °C
MDL Number: MFCD11848554
CAS Number: 178896-77-0

Order Now

Specification Test Results

Melting point 105-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR