N-Methoxy-N-methyl-1-[4-nitro-2-(trifluoromethyl)phenyl]-3-piperidinecarboxamide ≥95%
Supplier: Matrix Scientific
046794-500MG
046794-1G
101877-038EA
375.38
USD
101877-038
101877-040
N-Methoxy-N-methyl-1-[4-nitro-2-(trifluoromethyl)phenyl]-3-piperidinecarboxamide ≥95%
N-Methoxy-N-methyl-1-[4-nitro-2-(trifluoromethyl)phenyl]-3-piperidinecarboxamide
Hazard: Irritant
Formula:
C₁₅H₁₈F₃N₃O₄ MW: 361.32 g/mol Melting Pt: 58…60 °C |
MDL Number:
MFCD12498689 CAS Number: 1171918-79-8 |
Specification Test Results
Melting point | 58-60°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...