Ethyl-1-[4-nitro-2-(trifluoromethyl)phenyl]-3-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

046790-500MG 046790-1G 046790-5G
101877-020EA 278.46 USD
101877-020 101877-022 101877-024
Ethyl-1-[4-nitro-2-(trifluoromethyl)phenyl]-3-piperidinecarboxylate ≥95%
Ethyl-1-[4-nitro-2-(trifluoromethyl)phenyl]-3-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₇F₃N₂O₄
Melting Pt: 59…61 °C
MDL Number: MFCD11042095
CAS Number: 927697-73-2

Order Now

Specification Test Results

Melting point 59-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR