2-{[4-(tert-Butoxycarbonyl)piperazino]carbonyl}cyclopropanecarboxylic acid ≥95%

Supplier: Matrix Scientific

046789-500MG 046789-1G
101877-016EA 921.54 USD
101877-016 101877-018
2-{[4-(tert-Butoxycarbonyl)piperazino]carbonyl}cyclopropanecarboxylic acid ≥95%
2-{[4-(tert-Butoxycarbonyl)piperazino]carbonyl}cyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₅
MW: 298.34 g/mol
Melting Pt: 193…194 °C
MDL Number: MFCD12025896
CAS Number: 1160474-45-2

Order Now

Specification Test Results

Melting point 193-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR