2-Cyano-2-methylpropanoic acid ≥95%

Supplier: Matrix Scientific

046787-500MG 046787-1G
101877-006EA 141.54 USD
101877-006 101877-008
2-Cyano-2-methylpropanoic acid ≥95%
2-Cyano-2-methylpropanoic acid

Hazard: Irritant

Formula: C₅H₇NO₂
Melting Pt: 153…155 °C
MDL Number: MFCD11100210
CAS Number: 22426-30-8

Order Now

Specification Test Results

Melting point 153-155°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR