2-Amino-5-bromo-3-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-(trifluoromethyl)pyridin-2-amine

046779-500MG 046779-1G
101876-976EA 927.69 USD
101876-976 101876-978
2-Amino-5-bromo-3-(trifluoromethyl)pyridine ≥95%
2-Amino-5-bromo-3-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₄BrF₃N₂
MW: 241.01 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD10758088
CAS Number: 79456-34-1

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR