Methyl-1-(aminomethyl)cyclopentanecarboxylate hydrochloride ≥97%

Supplier: Matrix Scientific

046778-500MG 046778-1G
101876-972EA 747.69 USD
101876-972 101876-974
Methyl-1-(aminomethyl)cyclopentanecarboxylate hydrochloride ≥97%
Methyl-1-(aminomethyl)cyclopentanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MW: 193.68 g/mol
Melting Pt: 175…176 °C
MDL Number: MFCD10758087
CAS Number: 1171925-49-7

Order Now

Specification Test Results

Melting point 175-176°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR