2-[4-(Trifluoromethyl)phenethyl]piperazine ≥95%
Supplier: Matrix Scientific
046775-500MG
046775-1G
101876-962EA
764.62
USD
101876-962
101876-964
2-[4-(Trifluoromethyl)phenethyl]piperazine ≥95%
2-[4-(Trifluoromethyl)phenethyl]piperazine
Hazard: Irritant
Formula:
C₁₃H₁₇F₃N₂ MW: 258.29 g/mol Melting Pt: 137…139 °C |
MDL Number:
MFCD11100209 CAS Number: 1033463-32-9 |
Specification Test Results
Melting point | 137-139°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...