Methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-hydroxy-3H-benzo[d]imidazole-5-carboxylate
046773-500MG
046773-1G
046773-5G
101876-950EA
118.46
USD
101876-950
101876-952
101876-954
Methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate ≥95%
Methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate
Hazard: Irritant
Formula:
C₉H₈N₂O₃ MW: 192.18 g/mol Melting Pt: 312…313 °C |
MDL Number:
MFCD08457979 CAS Number: 106429-57-6 |
Specification Test Results
Melting point | 312-313°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...