2-Chloro-N-[3-(1-pyrrolidinyl)propyl]benzenesulfonamide ≥95%
Supplier: Matrix Scientific
046770-500MG
046770-1G
101876-938EA
181.54
USD
101876-938
101876-940
2-Chloro-N-[3-(1-pyrrolidinyl)propyl]benzenesulfonamide ≥95%
2-Chloro-N-[3-(1-pyrrolidinyl)propyl]benzenesulfonamide
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₂S MW: 302.83 g/mol Melting Pt: 75…77 °C |
MDL Number:
MFCD02557237 CAS Number: 321721-53-3 |
Specification Test Results
Melting point | 75-77°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...