2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
046767-500MG
046767-1G
046767-5G
101876-924EA
175.38
USD
101876-924
101876-926
101876-928
2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid ≥95%
2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₂ MW: 190.20 g/mol Melting Pt: 122-123°C |
MDL Number:
MFCD00269632 CAS Number: 874605-59-1 |
Specification Test Results
Melting point | 122-123°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...