3-Chloro-5-(trifluoromethyl)-2-pyridinecarbohydrazide ≥90%

Supplier: Matrix Scientific

046765-500MG 046765-1G
101876-920EA 556.92 USD
101876-920 101876-922
3-Chloro-5-(trifluoromethyl)-2-pyridinecarbohydrazide ≥90%
3-Chloro-5-(trifluoromethyl)-2-pyridinecarbohydrazide

Hazard: Irritant

Formula: C₇H₅ClF₃N₃O
Melting Pt: 105…107 °C
MDL Number: MFCD10758083
CAS Number: 1033463-30-7

Order Now

Specification Test Results

Melting point 105-107°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR