7-Bromo-1H-indazole ≥95%
Supplier: Matrix Scientific
046762-1G
046762-5G
101876-910EA
60
USD
101876-910
101876-912
7-Bromo-1H-indazole ≥95%
7-Bromo-1H-indazole
Hazard: Irritant
Formula:
C₇H₅BrN₂ MW: 197.0300 g/mol Melting Pt: 123…125 °C |
MDL Number:
MFCD07371564 CAS Number: 53857-58-2 |
Specification Test Results
Melting point | 123-125°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...