2-(4-Chlorobenzyl)-4-methyl-1,3-thiazole-5-carboxylic acid ≥95%
Supplier: Matrix Scientific
046761-500MG
046761-1G
046761-5G
101876-904EA
556.92
USD
101876-904
101876-906
101876-908
2-(4-Chlorobenzyl)-4-methyl-1,3-thiazole-5-carboxylic acid ≥95%
2-(4-Chlorobenzyl)-4-methyl-1,3-thiazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₀ClNO₂S Melting Pt: 243…245 °C |
MDL Number:
MFCD08558314 CAS Number: 938001-17-3 |
Specification Test Results
Melting point | 243-245°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...