5-Amino-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrazole-4-carbonitrile ≥95%
Supplier: Matrix Scientific
046757-500MG
046757-1G
101876-888EA
278.46
USD
101876-888
101876-890
5-Amino-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrazole-4-carbonitrile
Hazard: Irritant
Formula:
C₁₀H₅ClF₃N₅ Melting Pt: 214…216 °C |
MDL Number:
MFCD10758081 CAS Number: 94038-76-3 |
Specification Test Results
Melting point | 214-216°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...