3-(3-Bromophenyl)-1H-pyrazole-4-carbonitrile ≥90%

Supplier: Matrix Scientific

046754-500MG 046754-1G
101876-874EA 538.46 USD
101876-874 101876-876
3-(3-Bromophenyl)-1H-pyrazole-4-carbonitrile ≥90%
3-(3-Bromophenyl)-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₀H₆BrN₃
Melting Pt: 136…138 °C
MDL Number: MFCD11100206
CAS Number: 1170378-39-8

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR