4-Amino-3-(3-bromophenyl)-1H-pyrazole ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(3-Bromophenyl)-1H-pyrazol-4-amine
046753-500MG
046753-1G
101876-870EA
556.92
USD
101876-870
101876-872
4-Amino-3-(3-bromophenyl)-1H-pyrazole ≥95%
4-Amino-3-(3-bromophenyl)-1H-pyrazole
Hazard: Irritant
Formula:
C₉H₈BrN₃ Melting Pt: 94…96 °C |
MDL Number:
MFCD11100205 CAS Number: 1171492-86-6 |
Specification Test Results
Melting point | 94-96°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...