5-(Chlorosulfonyl)-2-methylbenzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Chlorosulphonyl)-2-methylbenzoic acid

046751-1G 046751-5G
101876-862EA 60 USD
101876-862 101876-864
5-(Chlorosulfonyl)-2-methylbenzenecarboxylic acid ≥95%
5-(Chlorosulfonyl)-2-methylbenzenecarboxylic acid

Hazard: Irritant

Formula: C₈H₇ClO₄S
MW: 234.66 g/mol
Melting Pt: 152…154 °C
MDL Number: MFCD03989642
CAS Number: 89001-57-0

Order Now

Specification Test Results

Melting point 152-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR