1-[2-(4-Ethylanilino)-1,3-thiazol-5-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

046742-500MG 046742-1G 046742-5G
101876-816EA 556.92 USD
101876-816 101876-818 101876-820
1-[2-(4-Ethylanilino)-1,3-thiazol-5-yl]-1-ethanone ≥95%
1-[2-(4-Ethylanilino)-1,3-thiazol-5-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₃H₁₄N₂OS
Melting Pt: 148-150°C
MDL Number: MFCD09607947
CAS Number: 952183-58-3

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR