6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 1,6-Dihydro-6-oxo-1-[3-(trifluoromethyl)benzyl]pyridine-3-carboxaldehyde

046741-500MG 046741-1G
101876-812EA 556.92 USD
101876-812 101876-814
6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀F₃NO₂
MW: 281.24 g/mol
Melting Pt: 91…93 °C
MDL Number: MFCD09817436
CAS Number: 952183-57-2

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR