1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
Supplier: Matrix Scientific
046740-500MG
046740-1G
101876-808EA
556.92
USD
101876-808
101876-810
1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₁₃H₉Cl₂NO₂ MW: 282.13 g/mol Melting Pt: 144…146 °C |
MDL Number:
MFCD09817435 CAS Number: 952183-56-1 |
Specification Test Results
Melting point | 144-146°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...