1-(4-Methylbenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
1,6-Dihydro-1-(4-methylbenzyl)-6-oxopyridine-3-carboxaldehyde
046739-500MG
046739-1G
101876-804EA
556.92
USD
101876-804
101876-806
1-(4-Methylbenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
1-(4-Methylbenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₃NO₂ MW: 227.27 g/mol Melting Pt: 107…109 °C |
MDL Number:
MFCD09817434 CAS Number: 952183-55-0 |
Specification Test Results
Melting point | 107-109°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...