Methyl-3-(ethylamino)-4-nitrobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
046738-500MG
046738-1G
046738-5G
101876-798EA
278.46
USD
101876-798
101876-800
101876-802
Methyl-3-(ethylamino)-4-nitrobenzenecarboxylate ≥95%
Methyl-3-(ethylamino)-4-nitrobenzenecarboxylate
Hazard: Irritant
Formula:
C₁₀H₁₂N₂O₄ Melting Pt: 92…94 °C |
MDL Number:
MFCD09607946 CAS Number: 952183-54-9 |
Specification Test Results
Melting point | 92-94°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...