5-Acetyl-2-chloropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Acetyl-6-chloropyridine

046735-1G 046735-5G
101876-786EA 27.69 USD
101876-786 101876-788
5-Acetyl-2-chloropyridine ≥95%
5-Acetyl-2-chloropyridine

Hazard: Irritant

Formula: C₇H₆ClNO
MW: 155.5816 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD03407343
CAS Number: 55676-22-7

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR