5-Hydroxy-2(1H)-quinolinone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxyquinolin-2(1H)-one

046726-500MG 046726-1G
101876-750EA 747.69 USD
101876-750 101876-752
5-Hydroxy-2(1H)-quinolinone ≥95%
5-Hydroxy-2(1H)-quinolinone

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: >300 °C
MDL Number: MFCD02181132
CAS Number: 31570-97-5

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR