Ethyl-2-methyl-1,3-benzothiazole-6-carboxylate ≥95%
Supplier: Matrix Scientific
046723-500MG
046723-1G
101876-742EA
375.38
USD
101876-742
101876-744
Ethyl-2-methyl-1,3-benzothiazole-6-carboxylate ≥95%
Ethyl-2-methyl-1,3-benzothiazole-6-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂S Melting Pt: 67…69 °C |
MDL Number:
MFCD00226438 CAS Number: 103646-25-9 |
Specification Test Results
Melting point | 67-69°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...