2,3-Dihydro-1-benzofuran-5-sulfonamide ≥95%
Supplier: Matrix Scientific
046719-500MG
046719-1G
046719-5G
101876-720EA
375.38
USD
101876-720
101876-722
101876-724
2,3-Dihydro-1-benzofuran-5-sulfonamide ≥95%
2,3-Dihydro-1-benzofuran-5-sulfonamide
Hazard: Irritant
Formula:
C₈H₉NO₃S MW: 199.23 g/mol Melting Pt: 164…166 °C |
MDL Number:
MFCD03069322 CAS Number: 112894-47-0 |
Specification Test Results
Melting point | 164-166°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...