2-(4-Bromophenyl)imidazo[1,2-a]pyridine-8-carboxylic acid ≥95%

Supplier: Matrix Scientific

046717-500MG 046717-1G
101876-712EA 747.69 USD
101876-712 101876-714
2-(4-Bromophenyl)imidazo[1,2-a]pyridine-8-carboxylic acid ≥95%
2-(4-Bromophenyl)imidazo[1,2-a]pyridine-8-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₉BrN₂O₂
Melting Pt: 235-238°C
MDL Number: MFCD09864798
CAS Number: 133427-42-6

Order Now

Specification Test Results

Melting point 235-238°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR