3-Oxo-3-(2-pyridinyl)propanenitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Oxo-3-(pyridin-2-yl)propanenitrile

046716-1G 046716-5G
101876-708EA 123.08 USD
101876-708 101876-710
3-Oxo-3-(2-pyridinyl)propanenitrile ≥98%
3-Oxo-3-(2-pyridinyl)propanenitrile

Hazard: Irritant/Refrigerate

Formula: C₈H₆N₂O
MW: 146.15 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD06656806
CAS Number: 54123-21-6

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR