3-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-5-ylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-[3-(trifluoromethyl)phenyl]-1H-pyrazole

046715-500MG 046715-1G 046715-5G
101876-702EA 375.38 USD
101876-702 101876-704 101876-706
3-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-5-ylamine ≥95%
3-[3-(Trifluoromethyl)phenyl]-1H-pyrazol-5-ylamine

Hazard: Irritant

Formula: C₁₀H₈F₃N₃
MW: 227.19 g/mol
Melting Pt: 130…132 °C
MDL Number: MFCD04110829
CAS Number: 209224-91-9

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR