5-Amino-3-(3-(trifluoromethyl)phenyl)isoxazole ≥95%
Supplier: Matrix Scientific
046714-500MG
046714-1G
046714-5G
101876-696EA
375.38
USD
101876-696
101876-698
101876-700
5-Amino-3-(3-(trifluoromethyl)phenyl)isoxazole ≥95%
5-Amino-3-(3-(trifluoromethyl)phenyl)isoxazole
Hazard: Irritant
Formula:
C₁₀H₇F₃N₂O Melting Pt: 91-93°C |
MDL Number:
MFCD07787059 CAS Number: 82360-94-9 |
Specification Test Results
Melting point | 91-93°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...