2,3-Dihydro-1-benzofuran-5-sulfonoylchloride ≥95%

Supplier: Matrix Scientific

046713-500MG 046713-1G 046713-5G
101876-690EA 278.46 USD
101876-690 101876-692 101876-694
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride ≥95%
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride

Hazard: Irritant

Formula: C₈H₇ClO₃S
MW: 218.66 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD02677703
CAS Number: 115010-11-2

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR