3-Oxo-3-[4-(trifluoromethyl)phenyl]propanenitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)benzoylacetonitrile

046711-500MG 046711-1G
101876-680EA 278.46 USD
101876-680 101876-682
3-Oxo-3-[4-(trifluoromethyl)phenyl]propanenitrile ≥95%
3-Oxo-3-[4-(trifluoromethyl)phenyl]propanenitrile

Hazard: Irritant

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
Melting Pt: 43…45 °C
MDL Number: MFCD02260793
CAS Number: 71682-94-5

Order Now

Specification Test Results

Melting point 43-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR