3-Oxo-3-pyridin-3-ylpropionitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Oxo-3-(3-pyridyl)propanenitrile

046707-1G 046707-5G
101876-660EA 140 USD
101876-660 102833-144
3-Oxo-3-pyridin-3-ylpropionitrile ≥97%
3-Oxo-3-pyridin-3-ylpropionitrile

Hazard: Irritant/Store in Freezer

Formula: C8H6N2O
MW: 146.150 g/mol
Melting Pt: 79…81 °C
MDL Number: MFCD08282797
CAS Number: 30510-18-0

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR