Ethyl-2-chloro-5-cyano-6-methylnicotinate ≥95%
Supplier: Matrix Scientific
046706-500MG
046706-1G
046706-5G
101876-652EA
66.15
USD
101876-652
101876-654
101876-656
Ethyl-2-chloro-5-cyano-6-methylnicotinate ≥95%
Ethyl-2-chloro-5-cyano-6-methylnicotinate
Hazard: Irritant
Formula:
C₁₀H₉ClN₂O₂ Melting Pt: 49…51 °C |
MDL Number:
MFCD01565859 CAS Number: 75894-43-8 |
Specification Test Results
Melting point | 49-51°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...