1-(4-Bromophenyl)tetrahydro-2H-imidazol-2-one ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-Bromophenyl)imidazolidin-2-one
046705-500MG
046705-1G
046705-5G
101876-646EA
126.15
USD
101876-646
101876-648
101876-650
1-(4-Bromophenyl)tetrahydro-2H-imidazol-2-one ≥95%
1-(4-Bromophenyl)tetrahydro-2H-imidazol-2-one
Hazard: Irritant
Formula:
C₉H₉BrN₂O MW: 241.09 g/mol Melting Pt: 189…191 °C |
MDL Number:
MFCD08444010 CAS Number: 530081-14-2 |
Specification Test Results
Melting point | 189-191°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...